5-(2-Fluoro-3-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55741-1G 55741-25G 55741-500MG 55741-5G
76933-912EA 352.22 USD
76933-912 76933-914 76933-916 76933-918
5-(2-Fluoro-3-methylphenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-3-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂S
MW: 208.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564321
CAS Number: 1861620-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR