5-Ethyl-2-fluoro-3-formylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

55703-1G 55703-500MG 55703-5G
76933-900EA 792.5 USD
76933-900 76933-902 76933-904
5-Ethyl-2-fluoro-3-formylbenzoic acid ≥97%
5-Ethyl-2-fluoro-3-formylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD34530434
CAS Number: 2710669-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR