5-(5-Fluoro-2-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55681-1G 55681-25G 55681-500MG 55681-5G
76933-884EA 441.34 USD
76933-884 76933-886 76933-888 76933-890
5-(5-Fluoro-2-methylphenyl)thiazol-2-amine ≥97%
5-(5-Fluoro-2-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂S
MW: 208.255 g/mol
Storage Temperature: Ambient
CAS Number: 1179718-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR