5-(3-Fluoro-2-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55630-1G 55630-25G 55630-500MG 55630-5G
76933-870EA 428.61 USD
76933-870 76933-872 76933-874 76933-876
5-(3-Fluoro-2-methylphenyl)thiazol-2-amine ≥97%
5-(3-Fluoro-2-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂S
MW: 208.255 g/mol
Storage Temperature: Ambient
CAS Number: 1550534-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR