5-(4-Fluoro-2-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55626-1G 55626-500MG 55626-5G
76933-856EA 464.68 USD
76933-856 76933-858 76933-860
5-(4-Fluoro-2-methylphenyl)thiazol-2-amine ≥97%
5-(4-Fluoro-2-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂S
MW: 208.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12779874
CAS Number: 1179232-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR