5-(4-Bromo-3-chlorophenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

55600-1G 55600-25G 55600-500MG 55600-5G
76933-842EA 436.03 USD
76933-842 76933-844 76933-846 76933-848
5-(4-Bromo-3-chlorophenyl)thiazol-2-amine ≥95%
5-(4-Bromo-3-chlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆BrClN₂S
MW: 289.579 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564312
CAS Number: 1695166-75-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR