5-(4-Bromo-3-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55515-1G 55515-25G 55515-500MG 55515-5G
76933-826EA 434.97 USD
76933-826 76933-828 76933-830 76933-832
5-(4-Bromo-3-fluorophenyl)thiazol-2-amine ≥97%
5-(4-Bromo-3-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆BrFN₂S
MW: 273.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564304
CAS Number: 1602832-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR