5-(3-Chloro-4-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55505-1G 55505-25G 55505-500MG 55505-5G
76933-812EA 405.27 USD
76933-812 76933-814 76933-816 76933-818
5-(3-Chloro-4-fluorophenyl)thiazol-2-amine ≥97%
5-(3-Chloro-4-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆ClFN₂S
MW: 228.674 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24221361
CAS Number: 1546702-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR