5-(2-(Methoxymethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55435-1G 55435-500MG 55435-5G
76933-774EA 695.96 USD
76933-774 76933-776 76933-778
5-(2-(Methoxymethyl)phenyl)thiazol-2-amine ≥97%
5-(2-(Methoxymethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂OS
MW: 220.291 g/mol
Storage Temperature: Ambient
CAS Number: 1600673-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR