5-(2-(Methylthio)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55413-1G 55413-25G 55413-500MG 55413-5G
76933-760EA 334.19 USD
76933-760 76933-762 76933-764 76933-766
5-(2-(Methylthio)phenyl)thiazol-2-amine ≥97%
5-(2-(Methylthio)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂S₂
MW: 222.33 g/mol
Storage Temperature: Ambient
CAS Number: 2109998-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR