5-(3-Ethoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55321-1G 55321-25G 55321-500MG 55321-5G
76933-746EA 396.78 USD
76933-746 76933-748 76933-750 76933-752
5-(3-Ethoxyphenyl)thiazol-2-amine ≥97%
5-(3-Ethoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂OS
Storage Temperature: Ambient
CAS Number: 1514725-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR