5-(2-Chloro-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55167-1G 55167-500MG 55167-5G
76933-666EA 530.45 USD
76933-666 76933-668 76933-670
5-(2-Chloro-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(2-Chloro-5-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆ClF₃N₂S
MW: 278.681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530360
CAS Number: 2734776-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR