5-(3,5-Bis(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55165-1G 55165-25G 55165-500MG 55165-5G
76933-652EA 376.62 USD
76933-652 76933-654 76933-656 76933-658
5-(3,5-Bis(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(3,5-Bis(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₆F₆N₂S
MW: 312.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530358
CAS Number: 1770153-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR