5-(2-Fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55154-1G 55154-25G 55154-500MG 55154-5G
76933-638EA 381.93 USD
76933-638 76933-640 76933-642 76933-644
5-(2-Fluorophenyl)thiazol-2-amine ≥97%
5-(2-Fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₇FN₂S
MW: 194.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846733
CAS Number: 1025927-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR