5-(3-(Methoxymethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55149-1G 55149-500MG 55149-5G
76933-626EA 589.87 USD
76933-626 76933-628 76933-630
5-(3-(Methoxymethyl)phenyl)thiazol-2-amine ≥97%
5-(3-(Methoxymethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂OS
MW: 220.291 g/mol
Storage Temperature: Ambient
CAS Number: 1601157-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR