5-(4-Isobutylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55135-1G 55135-25G 55135-500MG 55135-5G
76933-612EA 396.78 USD
76933-612 76933-614 76933-616 76933-618
5-(4-Isobutylphenyl)thiazol-2-amine ≥97%
5-(4-Isobutylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂S
Storage Temperature: Ambient
CAS Number: 1179145-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR