5-(3-(Methylthio)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55127-1G 55127-500MG 55127-5G
76933-598EA 461.5 USD
76933-598 76933-600 76933-602
5-(3-(Methylthio)phenyl)thiazol-2-amine ≥97%
5-(3-(Methylthio)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂S₂
Storage Temperature: Ambient
CAS Number: 2150253-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR