5-(4-(Methoxymethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55088-1G 55088-500MG 55088-5G
76933-556EA 589.87 USD
76933-556 76933-558 76933-560
5-(4-(Methoxymethyl)phenyl)thiazol-2-amine ≥97%
5-(4-(Methoxymethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂OS
MW: 220.291 g/mol
Storage Temperature: Ambient
CAS Number: 1595959-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR