5-(3,4-Dimethylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55062-1G 55062-25G 55062-500MG 55062-5G
76933-542EA 375.56 USD
76933-542 76933-544 76933-546 76933-548
5-(3,4-Dimethylphenyl)thiazol-2-amine ≥97%
5-(3,4-Dimethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂S
MW: 204.291 g/mol
Storage Temperature: Ambient
CAS Number: 1178218-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR