5-(3-Fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54987-1G 54987-500MG 54987-5G
76933-506EA 471.04 USD
76933-506 76933-508 76933-510
5-(3-Fluorophenyl)thiazol-2-amine ≥97%
5-(3-Fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₇FN₂S
MW: 194.229 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846734
CAS Number: 438585-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR