5-(4-Chlorophenyl)thiazol-2-amine ≥97% research grade

Supplier: AOBChem USA

Warning

54980-1G 54980-500MG 54980-5G
76933-492EA 492.26 USD
76933-492 76933-494 76933-496
5-(4-Chlorophenyl)thiazol-2-amine ≥97% research grade
5-(4-Chlorophenyl)thiazol-2-amine
Formula: C₉H₇ClN₂S
MW: 210.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12779839
CAS Number: 73040-66-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR