4-Bromo-2-fluoro-5-methylbenzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
54912-25G
54912-5G
76933-466EA
219.61
USD
76933-466
76933-468
4-Bromo-2-fluoro-5-methylbenzoic acid ≥97%
4-Bromo-2-fluoro-5-methylbenzoic acid
Formula:
C₈H₆BrFO₂ Storage Temperature: Ambient |
MDL Number:
MFCD08458245 CAS Number: 415965-24-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...