5-(4-(Trifluoromethoxy)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54853-1G 54853-500MG 54853-5G
76933-454EA 500.75 USD
76933-454 76933-456 76933-458
5-(4-(Trifluoromethoxy)phenyl)thiazol-2-amine ≥97%
5-(4-(Trifluoromethoxy)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₇F₃N₂OS
MW: 260.236 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12859692
CAS Number: 1183699-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR