5-(5-Bromo-2-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54849-1G 54849-25G 54849-500MG 54849-5G
76933-438EA 396.78 USD
76933-438 76933-440 76933-442 76933-444
5-(5-Bromo-2-fluorophenyl)thiazol-2-amine ≥97%
5-(5-Bromo-2-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆BrFN₂S
MW: 273.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23759019
CAS Number: 1537952-88-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR