5-Bromo-2,3-dimethoxybenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

54707-1G 54707-25G 54707-5G
76933-424EA 111.4 USD
76933-424 76933-426 76933-428
5-Bromo-2,3-dimethoxybenzaldehyde ≥95%
5-Bromo-2,3-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02611624
CAS Number: 71295-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR