5-(2,6-Dimethylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54302-1G 54302-500MG 54302-5G
76933-384EA 471.04 USD
76933-384 76933-386 76933-388
5-(2,6-Dimethylphenyl)thiazol-2-amine ≥97%
5-(2,6-Dimethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂S
MW: 204.291 g/mol
Storage Temperature: Ambient
CAS Number: 1598080-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR