5-(3-Methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

54292-1G 54292-500MG 54292-5G
76933-370EA 471.04 USD
76933-370 76933-372 76933-374
5-(3-Methoxyphenyl)thiazol-2-amine ≥97%
5-(3-Methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂OS
Storage Temperature: Ambient
MDL Number: MFCD12824100
CAS Number: 1183012-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR