4-Chloro-2-(methoxymethoxy)-1-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

54280-25G 54280-5G
76933-360EA 204.76 USD
76933-360 76933-362
4-Chloro-2-(methoxymethoxy)-1-methylbenzene ≥97%
4-Chloro-2-(methoxymethoxy)-1-methylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂
MW: 186.635 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD21285563
CAS Number: 1468717-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR