5-Chloro-2-methoxy-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

53639-1G 53639-500MG 53639-5G
76933-306EA 629.12 USD
76933-306 76933-308 76933-310
5-Chloro-2-methoxy-4-methylbenzoic acid ≥97%
5-Chloro-2-methoxy-4-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MW: 200.619 g/mol
Storage Temperature: Ambient
CAS Number: 28478-41-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR