5-Bromo-2-chloro-6-methylnicotinaldehyde ≥95%

Supplier: AOBChem USA

Warning

53532-1G 53532-25G 53532-500MG 53532-5G
76933-290EA 126.25 USD
76933-290 76933-292 76933-294 76933-296
5-Bromo-2-chloro-6-methylnicotinaldehyde ≥95%
5-Bromo-2-chloro-6-methylnicotinaldehyde

Precaución: for Research Use Only

Formula: C₇H₅BrClNO
Storage Temperature: Refrigerator
CAS Number: 1935331-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR