5-Bromo-[1,1'-biphenyl]-2-ol ≥97%

Supplier: AOBChem USA

Warning

53072-1G 53072-25G 53072-500MG 53072-5G
76933-232EA 334.19 USD
76933-232 76933-234 76933-236 76933-238
5-Bromo-[1,1'-biphenyl]-2-ol ≥97%
5-Bromo-[1,1'-biphenyl]-2-ol

Precaución: for Research Use Only

Formula: C₁₂H₉BrO
MW: 249.103 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18459194
CAS Number: 16434-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR