4-Methylbenzo[d][1,3]dioxole-5-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

53005-1G 53005-25G 53005-500MG 53005-5G
76933-212EA 237.65 USD
76933-212 76933-214 76933-216 76933-218
4-Methylbenzo[d][1,3]dioxole-5-carboxylic acid ≥97%
4-Methylbenzo[d][1,3]dioxole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈O₄
Storage Temperature: Ambient
CAS Number: 162506-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR