5-Chloro-[1,1'-biphenyl]-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

52939-1G 52939-25G 52939-500MG 52939-5G
76933-198EA 215.37 USD
76933-198 76933-200 76933-202 76933-204
5-Chloro-[1,1'-biphenyl]-3-carboxylic acid ≥97%
5-Chloro-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₉ClO₂
MW: 232.662 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11577111
CAS Number: 1082283-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR