4-Bromo-2-fluorostyrene ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 4-Bromo-2-fluoro-1-vinylbenzene, 4-Bromo-1-ethenyl-2-fluorobenzene

52671-1G 52671-25G 52671-500MG 52671-5G
76933-172EA 200.52 USD
76933-172 76933-174 76933-176 76933-178
4-Bromo-2-fluorostyrene ≥97%
4-Bromo-2-fluorostyrene
Formula: C₈H₆BrF
MW: 201.036 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09907860
CAS Number: 627463-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR