4-Formyl-2,5-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52561-1G 52561-500MG 52561-5G
76933-150EA 560.16 USD
76933-150 76933-152 76933-154
4-Formyl-2,5-dimethylbenzoic acid ≥97%
4-Formyl-2,5-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MW: 178.185 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31432042
CAS Number: 477735-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR