5-Chloro-3,3-difluoroindolin-2-one ≥97%

Supplier: AOBChem USA

Warning

52312-1G 52312-25G 52312-500MG 52312-5G
76933-118EA 399.96 USD
76933-118 76933-120 76933-122 76933-124
5-Chloro-3,3-difluoroindolin-2-one ≥97%
5-Chloro-3,3-difluoroindolin-2-one

Precaución: for Research Use Only

Formula: C₈H₄ClF₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD22547888
CAS Number: 197067-32-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR