4-Butoxy-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52279-1G 52279-25G 52279-500MG 52279-5G
76933-102EA 257.8 USD
76933-102 76933-104 76933-106 76933-108
4-Butoxy-2-methylbenzoic acid ≥97%
4-Butoxy-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₃
Storage Temperature: Refrigerator
MDL Number: MFCD11226977
CAS Number: 175153-56-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR