4-Fluoro-2-methoxybenzamide ≥97% research grade

Supplier: AOBChem USA

Warning

51696-1G 51696-25G 51696-5G
76933-060EA 62.6 USD
76933-060 76933-062 76933-064
4-Fluoro-2-methoxybenzamide ≥97% research grade
4-Fluoro-2-methoxybenzamide
Formula: C₈H₈FNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD06409084
CAS Number: 874804-07-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR