5-Bromo-3-chloropicolinaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

50785-1G 50785-25G 50785-500MG 50785-5G
76932-992EA 129.43 USD
76932-992 76932-994 76932-996 76932-998
5-Bromo-3-chloropicolinaldehyde ≥97% research grade
5-Bromo-3-chloropicolinaldehyde
Formula: C₆H₃BrClNO
Storage Temperature: Refrigerator
MDL Number: MFCD08277307
CAS Number: 885168-04-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR