4-Bromo-3-chloro-5-fluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

50168-1G 50168-25G 50168-500MG 50168-5G
76932-976EA 439.22 USD
76932-976 76932-978 76932-980 76932-982
4-Bromo-3-chloro-5-fluorobenzaldehyde ≥97%
4-Bromo-3-chloro-5-fluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrClFO
MW: 237.454 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28126801
CAS Number: 1781010-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR