4-Bromo-3,5-dichlorophenol ≥97%

Supplier: AOBChem USA

Warning

50037-1G 50037-25G 50037-500MG 50037-5G
76932-960EA 203.7 USD
76932-960 76932-962 76932-964 76932-966
4-Bromo-3,5-dichlorophenol ≥97%
4-Bromo-3,5-dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂O
MW: 241.897 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD1830114
CAS Number: 1940-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR