5-Bromo-2-chloro-3-nitrotoluene ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-chloro-1-methyl-3-nitrobenzene

49628-1G 49628-25G 49628-5G
76932-930EA 62.6 USD
76932-930 76932-932 76932-934
5-Bromo-2-chloro-3-nitrotoluene ≥97% research grade
5-Bromo-2-chloro-3-nitrotoluene
Formula: C₇H₅BrClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11846031
CAS Number: 1160573-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR