5-(Benzyloxy)-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

49440-1G 49440-25G 49440-500MG 49440-5G
76932-914EA 203.7 USD
76932-914 76932-916 76932-918 76932-920
5-(Benzyloxy)-2-methoxybenzaldehyde ≥97%
5-(Benzyloxy)-2-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00436291
CAS Number: 52329-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR