5-Bromo-2-(4-methylphenoxy)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 5-Bromo-2-(p-tolyloxy)pyridine

49190-1G 49190-25G 49190-500MG 49190-5G
76932-900EA 239.77 USD
76932-900 76932-902 76932-904 76932-906
5-Bromo-2-(4-methylphenoxy)pyridine ≥97%
5-Bromo-2-(4-methylphenoxy)pyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀BrNO
MW: 264.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14652098
CAS Number: 1248915-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR