5-Bromo-2-iodo-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

49166-1G 49166-25G 49166-500MG 49166-5G
76932-886EA 350.1 USD
76932-886 76932-888 76932-890 76932-892
5-Bromo-2-iodo-3-methylbenzoic acid ≥97%
5-Bromo-2-iodo-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrIO₂
Storage Temperature: Refrigerator
MDL Number: MFCD18379817
CAS Number: 1263376-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR