5-Chloro-2-(methoxymethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

49112-1G 49112-25G 49112-500MG 49112-5G
76932-872EA 233.4 USD
76932-872 76932-874 76932-876 76932-878
5-Chloro-2-(methoxymethoxy)benzaldehyde ≥95%
5-Chloro-2-(methoxymethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MW: 200.619 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21195358
CAS Number: 115560-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR