5-Bromo-2-fluoro-1-iodo-3-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

48937-1G 48937-25G 48937-500MG 48937-5G
76932-856EA 228.1 USD
76932-856 76932-858 76932-860 76932-862
5-Bromo-2-fluoro-1-iodo-3-methoxybenzene ≥97%
5-Bromo-2-fluoro-1-iodo-3-methoxybenzene

Precaución: for Research Use Only

Formula: C₇H₅BrFIO
MW: 330.921 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD32902784
CAS Number: 2386783-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR