4-Bromo-3-ethoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

48755-1G 48755-25G 48755-500MG 48755-5G
76932-832EA 381.93 USD
76932-832 76932-834 76932-836 76932-838
4-Bromo-3-ethoxybenzoic acid ≥97%
4-Bromo-3-ethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
MW: 245.07 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09753722
CAS Number: 933671-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR