4-Fluoro-3-(methoxymethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

48500-1G 48500-25G 48500-500MG 48500-5G
76932-812EA 332.07 USD
76932-812 76932-814 76932-816 76932-818
4-Fluoro-3-(methoxymethyl)phenylboronic acid ≥97%
4-Fluoro-3-(methoxymethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BFO₃
MW: 183.973 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28147726
CAS Number: 337536-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR