4-Bromo-2-isopropoxy-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

48452-25G 48452-5G
76932-784EA 559.1 USD
76932-784 76932-786
4-Bromo-2-isopropoxy-1-methoxybenzene ≥97%
4-Bromo-2-isopropoxy-1-methoxybenzene
Formula: C₁₀H₁₃BrO₂
Storage Temperature: Ambient
MDL Number: MFCD10699696
CAS Number: 462092-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR